Learn More
Waters Corp Column Kit, C18, 2.5 µm, 130 Å, 4.6 x 100 mm
Supplier:  Waters Corp 176003075
Encompass_Preferred
This CSH C18 column kit features particles with low, defined positive charges, ensuring excellent peptide peak shape with FA- or TFA-containing mobile phases. Suitable for LC or LC-MS, it provides exceptional separation performance and throughput for HPLC, facilitating UPLC adoption.
  • Provides excellent peak shape for peptides using FA- or TFA-containing mobile phases.
  • Suitable for LC or LC-MS applications; minimal dependence on strong ion pairing agents.
  • Packed in ultra-low dispersion eXtended Performance (XP) hardware.
  • Includes CSH C18 column and Cytochrome C Digestion Standard for benchmarking.
  • Features trifunctional C18 ligand, fully end-capped, bonded to Charged Surface Hybrid (CSH) particles.
  • Accepts greater peptide mass loads, useful for detecting low-level impurities.
  • 130Å pore size best suited for compounds less than 10,000 Dalton.
Catalog No. 50-239-1096