Learn More
Waters Corp XSelect Peptide CSH C18 Column Kit, 130 Å, 3.5 μm, 2.1 mm x 10 mm
Catalog No. 502391110 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters XSelect Peptide CSH C18 columns are designed for peptide mapping and analysis, offering excellent peak shape and high efficiency. These columns utilize Charged Surface Hybrid (CSH) Technology with C18 bonding and end-capping, ensuring reproducible performance for a wide range of peptide applications.
  • Qualified for peptide mapping, ensuring excellent, reproducible performance.
  • Same particle technology as other UPLC and HPLC columns for seamless transferability between system platforms.
  • CSH technology provides excellent peak shape, high efficiency, and loading capacity for basic compounds in acidic mobile phases.
  • OBD™ difference ensures uniform bed density and predictable scale-up performance.
  • Includes guidance for optimizing HPLC systems for narrow-bore columns to improve sensitivity and resolution in LC-MS/MS systems.
Catalog No. 50-239-1110 Supplier Waters Corp Supplier No. 176003081
Waters XSelect Peptide CSH C18 columns are designed for peptide mapping and analysis, offering excellent peak shape and high efficiency. These columns utilize Charged Surface Hybrid (CSH) Technology with C18 bonding and end-capping, ensuring reproducible performance for a wide range of peptide applications.
  • Qualified for peptide mapping, ensuring excellent, reproducible performance.
  • Same particle technology as other UPLC and HPLC columns for seamless transferability between system platforms.
  • CSH technology provides excellent peak shape, high efficiency, and loading capacity for basic compounds in acidic mobile phases.
  • OBD™ difference ensures uniform bed density and predictable scale-up performance.
  • Includes guidance for optimizing HPLC systems for narrow-bore columns to improve sensitivity and resolution in LC-MS/MS systems.