Learn More
Waters Corp Column Kit, Peptide CSH C18, 130Å, 3.5 μm
Catalog No. 502391441 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column kit features 3.5 μm CSH130 C18 particles designed for excellent peptide peak shape in FA- or TFA-containing mobile phases. It shows minimal dependence on strong ion pairing agents, making it ideal for LC or LC-MS applications.
  • Optimized peak shape and capacity for optical and LC-MS detection.
  • Enhanced mass load acceptance for detecting low-level impurities.
  • 130Å pore size, suitable for peptides under 10,000 Daltons.
  • Ensures consistent performance through rigorous quality control testing.
  • Includes a CSH130 C18 column and Cytochrome C Digestion Standard.
  • Certificate of analysis verifies quality and performance.
Catalog No. 50-239-1441 Supplier Waters Corp Supplier No. 176003078
This column kit features 3.5 μm CSH130 C18 particles designed for excellent peptide peak shape in FA- or TFA-containing mobile phases. It shows minimal dependence on strong ion pairing agents, making it ideal for LC or LC-MS applications.
  • Optimized peak shape and capacity for optical and LC-MS detection.
  • Enhanced mass load acceptance for detecting low-level impurities.
  • 130Å pore size, suitable for peptides under 10,000 Daltons.
  • Ensures consistent performance through rigorous quality control testing.
  • Includes a CSH130 C18 column and Cytochrome C Digestion Standard.
  • Certificate of analysis verifies quality and performance.