Learn More
Waters Corp Waters Column, CSH C18, 3.5 μm, 2.1 mm x 50 mm, 130Å
Supplier:  Waters Corp 186006950
Encompass_Preferred
This XSelect Peptide CSH C18 Column offers high-performance, high-selectivity for reversed-phase liquid chromatography of peptides, retaining polar and non-polar compounds on a 130Å, 3.5μm hybrid silica substrate.
  • High-performance, high-selectivity reversed-phase peptide chromatography.
  • 130Å, 3.5 μm sorbent bed on hybrid silica substrate.
  • Retains polar and non-polar compounds efficiently.
  • Compatible with major liquid chromatography systems.
  • Superior selectivity for complex peptide separation.
  • Convenient single-use format.
  • Enhances basic compound retention via embedded polar groups.
  • Ensures excellent peptide peak shape with low positive charge concentration.
  • Ideal for LC/LC-MS; minimal dependence on strong ion pairing agents.
  • Improves analytical performance.
Catalog No. 50-203-7778