Learn More
Waters Corp Guard Column, CSH C18, 5 μm, 130 Å, 30 mm X 10 mm
Supplier: Waters Corp 186007206
This guard column protects HPLC columns from particulate contaminants and absorptive compounds in mobile phase streams and samples. It integrates seamlessly into existing workflows, prolonging the analytical column's life. Packed with the same stationary phase and inner diameter as the analytical column, it helps mitigate backpressures.
- Superior column protection: Extends HPLC column lifespan.
- Excellent peak shape for peptides.
- LC and LC-MS compatibility.
- Seamless workflow integration.
- Consistent stationary phase.
- Mitigates backpressure.
- Cost-effective maintenance.
- Durable performance: Operates from 1 to 11 pH, withstands 6000 psi (415 Bar).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.