Learn More
Waters Corp XSelect Method Validation Kit, 130 Å, 2.5 μm, 2.1 mm x 100 mm, 3 pk
Catalog No. 502391230 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters CSH130 C18 based particles incorporate a low, carefully defined concentration of positive charges, contributing to excellent peak shape for peptides using FA or TFA mobile phases. Minimal dependence on strong ion pairing agents makes it suitable for LC or LC-MS applications. XSelect CSH130 C18 2.5 μm XP columns are packed in ultra-low dispersion hardware, facilitating exceptional separation performance, robustness, throughput for HPLC assays, and smooth transition to UPLC.
  • Includes three columns, each packed with a different batch of sorbent.
  • Contains one vial of Cytochrome C Digestion Standard.
  • Standard allows for immediate benchmarking of the column upon receipt.
  • Each batch of digestion standard is prepared under strict quality control criteria, confirmed by an included certificate of analysis.
Catalog No. 50-239-1230 Supplier Waters Corp Supplier No. 176003073
The Waters CSH130 C18 based particles incorporate a low, carefully defined concentration of positive charges, contributing to excellent peak shape for peptides using FA or TFA mobile phases. Minimal dependence on strong ion pairing agents makes it suitable for LC or LC-MS applications. XSelect CSH130 C18 2.5 μm XP columns are packed in ultra-low dispersion hardware, facilitating exceptional separation performance, robustness, throughput for HPLC assays, and smooth transition to UPLC.
  • Includes three columns, each packed with a different batch of sorbent.
  • Contains one vial of Cytochrome C Digestion Standard.
  • Standard allows for immediate benchmarking of the column upon receipt.
  • Each batch of digestion standard is prepared under strict quality control criteria, confirmed by an included certificate of analysis.