Learn More
Waters Corp WATERS CORPORATION Method Validation Kit, XSelect Peptide CSH C18, 4.6 x 100 mm, 3.5 μm
Supplier:  Waters Corp 186006959
Encompass_Preferred
This method validation kit provides essential components for rigorous peptide separations. It includes multiple columns and a digestion standard for consistent and reliable results.
  • Includes three columns from three different batches of CSH130 C18 material.
  • Includes one vial of Cytochrome c Digestion Standard.
  • Features column dimensions of 4.6 × 100 mm.
  • Designed with a particle size of 3.5 μm.
  • Offers a pore size of 130 Å.
Catalog No. 50-239-2524