Learn More
Waters Corp XSelect Peptide CSH C18 Column, Method Validation Kit, 4.6 mm × 100 mm, 3.5 μm
Supplier:  Waters Corp 176003086
Encompass_Preferred
The XSelect Peptide CSH C18 Column Kit is a method validation kit designed for superior peptide separations in LC or LC-MS applications. It features Charged Surface Hybrid particles that deliver excellent performance.
  • Charged Surface Hybrid particles for superior peptide separations in LC or LC-MS applications.
  • CSH Technology allows use with standard TFA-containing eluents or weaker acid modifiers.
  • Offers greater peptide mass loadability and outstanding peak capacities in HPLC or LC-MS.
  • QC tested for column-to-column reproducibility using a cytochrome c tryptic digest.
  • Suitable for peptide mapping, proteomics, and lab-scale purification.
  • Robust performance with little dependence on strong ion pairing agents.
  • Features stainless steel hardware.
  • Operates within a wide pH range of 1.0 to 11.0.
Catalog No. 50-239-2596