Learn More
Waters Corp Waters XSelect Peptide CSH C18 Prep Column, 130Å Pore, 5 μm Particle, 19 mm x 150 mm
Supplier: Waters Corp 186007021
XSelect CSH130 C18 5 μm particles offer excellent peak shape for peptides using FA- or TFA-containing mobile phases due to low, carefully-defined positive charges. Its performance has little dependence on strong ion pairing agents, making it ideal for LC or LC-MS applications.
- C18 chemistry
- Reversed phase separation mode
- pH range: 1 to 11
- Maximum pressure: 6000 psi (415 Bar)
- Endcapped
- Low silanol activity
- Molecular weight range: 1000 to 15000
- Spherical particle shape
- Particle size: 5 μm
- Pore size: 130 Å
- QC tested for peptide applications
- OBD Prep Column format
- Compatible with HPLC systems
- CSH particle technology
- Inner diameter: 19 mm
- Length: 150 mm
- Carbon load: 15 %
- One unit per package
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.