Learn More
Waters Corp XSelect Peptide CSH C18 OBD Column, 5 μm Particle Size
Supplier: Waters Corp 186007027
The XSelect Peptide CSH C18 OBD Column is designed for high-performance liquid chromatography. It features a particle size of 5 μm and dimensions of 50 x 100 mm, with a pore size of 130 Å.
- Specifically QC tested with a complex peptide mixture to help ensure column-to-column performance consistency.
- Dimensions of 50 x 100 mm.
- Particle size of 5 μm.
- Pore size of 130 Å.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.