Learn More
Waters Corp Waters XSelect OBD Prep Column, CSH C18, 5 μm, 130 Å, 10 mm x 100 mm
Catalog No. 502038464 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column incorporates a low and precisely defined concentration of positive charges. This design results in excellent peak shape for peptides when using either FA- or TFA-containing mobile phases. Its performance shows minimal dependence on strong ion pairing agents, making it suitable for LC or LC-MS applications.
  • Contains a low and carefully-defined concentration of positive charges.
  • Yields comparatively excellent peak shape for peptides using either FA- or TFA-containing mobile phases.
  • Performance exhibits little dependence on strong ion pairing agents.
  • Ideal for LC or LC-MS applications.
Catalog No. 50-203-8464 Supplier Waters Corp Supplier No. 186008265
This column incorporates a low and precisely defined concentration of positive charges. This design results in excellent peak shape for peptides when using either FA- or TFA-containing mobile phases. Its performance shows minimal dependence on strong ion pairing agents, making it suitable for LC or LC-MS applications.
  • Contains a low and carefully-defined concentration of positive charges.
  • Yields comparatively excellent peak shape for peptides using either FA- or TFA-containing mobile phases.
  • Performance exhibits little dependence on strong ion pairing agents.
  • Ideal for LC or LC-MS applications.