Learn More
Waters Corp WATERS CORPORATION Column, CSH130 C18, 130Å, 5 μm, 4.6 x 100 mm
Supplier:  Waters Corp 186007077
Encompass_Preferred
This column is engineered for lab-scale preparative chromatography, leveraging Charged Surface Hybrid (CSH) particles. The CSH technology provides a low-level positive charge, allowing effective use with both standard TFA-containing and weaker acid eluents, ensuring sharp, symmetrical peaks and minimal MS signal reduction.
  • Outstanding peak capacity for peptide and small protein separations (up to 10,000 daltons) in HPLC, UPLC®, or LC-MS, with 0.1% FA- or 0.1% TFA-containing eluents.
  • Excellent mass loading for complex peptide samples, ideal for peptide mapping, proteomics, and synthetic peptide analysis or purification.
  • Scalable chemistry from nano flow to lab-scale purification.
  • QC tested with a cytochrome c tryptic digest using 0.1% FA-containing eluents for consistent column-to-column reproducibility.
Catalog No. 50-203-7823