Learn More
Waters Corp Guard Cartridge, XSelect Peptide CSH C18, 130 Å, 3.5 μm, 4.6 x 20 mm
Supplier:  Waters Corp 186006958
Encompass_Preferred
This guard cartridge utilizes Charged Surface Hybrid (CSH) Technology to protect chromatography systems from contaminants. It provides excellent peak shape, high efficiency, and loading capacity for basic compounds, especially when acidic, low ionic strength mobile phases are used.
  • Features Charged Surface Hybrid (CSH) Technology.
  • Provides excellent peak shape, high efficiency, and high loading capacity for basic compounds.
  • Enables seamless transferability between HPLC and UPLC system platforms.
  • Manufactured in a cGMP, ISO 9001 certified facility using ultra-pure reagents.
  • Each batch is chromatographically tested to narrow specification ranges for reproducible performance.
  • Material produced from Peptide BEH C18, 130 Å particles with surface modification, C18 bonding, and end capping.
  • Compatible with preparative guards and holders.
Catalog No. 50-239-2656