Learn More
Waters Corp Waters XSelect Peptide CSH C18 Column, 2.5 μm, 2.1 x 150 mm
Catalog No. NC1819220 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These XSelect Peptide CSH C18 XP columns utilize CSH130 C18 particles with a low, precisely-defined concentration of positive charges, ensuring excellent peak shape for peptides. Designed for LC or LC-MS applications, they offer exceptional separation performance, robustness, and throughput for HPLC assays, facilitating a smooth transition to UPLC.
  • Charged surface hybrid (CSH) technology provides superior peak form and enhanced basic compound loading.
  • 2.5 μm extended performance (XP) columns offer exceptional separation performance and robustness.
  • Compatible with all HPLC, UPLC, and UHPLC technologies.
  • Withstands pressures up to 18,000 psi (1240 bar).
  • Usable within a pH range of 1 to 11, supporting a 15% carbon load.
  • Provides high peak capacity and lower retentivity for peptides with positive charges.
Catalog No. NC1819220 Supplier Waters Corp Supplier No. 186006943
These XSelect Peptide CSH C18 XP columns utilize CSH130 C18 particles with a low, precisely-defined concentration of positive charges, ensuring excellent peak shape for peptides. Designed for LC or LC-MS applications, they offer exceptional separation performance, robustness, and throughput for HPLC assays, facilitating a smooth transition to UPLC.
  • Charged surface hybrid (CSH) technology provides superior peak form and enhanced basic compound loading.
  • 2.5 μm extended performance (XP) columns offer exceptional separation performance and robustness.
  • Compatible with all HPLC, UPLC, and UHPLC technologies.
  • Withstands pressures up to 18,000 psi (1240 bar).
  • Usable within a pH range of 1 to 11, supporting a 15% carbon load.
  • Provides high peak capacity and lower retentivity for peptides with positive charges.