Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Column, C18, 5 μm, 2.1 mm x 100 mm, 100 Å

Catalog No. 502391024 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The XSelect Peptide HSS T3 Column offers silica-based selectivity and enhanced retention for hydrophilic peptides. Each batch of HSS T3, 100Å material is quality control tested with a tryptic digest of cytochrome c to meet stringent performance criteria.

  • C18 chemistry
  • Reversed-phase separation
  • Silica particle substrate
  • pH range: 2 to 8
  • 6000 psi (415 Bar) max pressure
  • Endcapped
  • T3 bonding technology
  • Particle size: 5 μm
  • Pore diameter: 100 Å
  • Inner diameter: 2.1 mm
  • Length: 100 mm
  • HPLC system compatible

Catalog No. 50-239-1024 Supplier Waters Corp Supplier No. 186008775
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The XSelect Peptide HSS T3 Column offers silica-based selectivity and enhanced retention for hydrophilic peptides. Each batch of HSS T3, 100Å material is quality control tested with a tryptic digest of cytochrome c to meet stringent performance criteria.

  • C18 chemistry
  • Reversed-phase separation
  • Silica particle substrate
  • pH range: 2 to 8
  • 6000 psi (415 Bar) max pressure
  • Endcapped
  • T3 bonding technology
  • Particle size: 5 μm
  • Pore diameter: 100 Å
  • Inner diameter: 2.1 mm
  • Length: 100 mm
  • HPLC system compatible

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.