Learn More
Waters Corp Peptide HSS T3 Column, 100Å, 2.5 μm, 4.6 mm x 50 mm
Catalog No. 502391039 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters XSelect Peptide HSS T3 Columns are designed for chromatographic separations requiring silica-based selectivities or enhanced retention of hydrophilic peptides. Each batch undergoes rigorous QC testing with a gradient separation of a tryptic digest of cytochrome c to ensure stringent performance criteria are met.
  • Designed for separations requiring silica-based selectivities.
  • Increased retention of hydrophilic peptides.
  • QC tested with gradient separation of a tryptic digest of cytochrome c.
  • Passes stringent performance criteria.
Catalog No. 50-239-1039 Supplier Waters Corp Supplier No. 186008762
Waters XSelect Peptide HSS T3 Columns are designed for chromatographic separations requiring silica-based selectivities or enhanced retention of hydrophilic peptides. Each batch undergoes rigorous QC testing with a gradient separation of a tryptic digest of cytochrome c to ensure stringent performance criteria are met.
  • Designed for separations requiring silica-based selectivities.
  • Increased retention of hydrophilic peptides.
  • QC tested with gradient separation of a tryptic digest of cytochrome c.
  • Passes stringent performance criteria.