Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp XSelect Peptide HSS T3 Column, 100 Å, 5 μm, 2.1 × 150 mm

Catalog No. 502391044 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The Waters XSelect Peptide HSS T3 Column is designed for efficient separation in various chromatographic applications. This 5 μm particle column features a 100 Å pore size and dimensions of 2.1 mm internal diameter by 150 mm length, optimized for peptide analysis.

  • Manufactured with High Strength Silica particle technology.
  • Supports operating pressures up to 5000 psi (400 bar or 40 MPa).
  • Stable within a pH range of 2 to 8.
  • Maximum recommended operating temperature of 45 °C.
  • Each batch undergoes rigorous quality control testing, including chromatographic analysis with diverse analytes and gradient separation of cytochrome c tryptic digest.
  • Produced in a cGMP, ISO 9001:2000 certified facility.
  • Equipped with standard endfittings, requiring a 0.130 in. ferrule depth.

Catalog No. 50-239-1044 Supplier Waters Corp Supplier No. 186008776
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Waters XSelect Peptide HSS T3 Column is designed for efficient separation in various chromatographic applications. This 5 μm particle column features a 100 Å pore size and dimensions of 2.1 mm internal diameter by 150 mm length, optimized for peptide analysis.

  • Manufactured with High Strength Silica particle technology.
  • Supports operating pressures up to 5000 psi (400 bar or 40 MPa).
  • Stable within a pH range of 2 to 8.
  • Maximum recommended operating temperature of 45 °C.
  • Each batch undergoes rigorous quality control testing, including chromatographic analysis with diverse analytes and gradient separation of cytochrome c tryptic digest.
  • Produced in a cGMP, ISO 9001:2000 certified facility.
  • Equipped with standard endfittings, requiring a 0.130 in. ferrule depth.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.