Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp XSelect Guard Column, 100Å pore, 2.5 μm particle, 2.1 mm × 5 mm

Catalog No. 502391861 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This guard column is optimized to protect and prolong analytical column lifetimes without compromising chromatographic performance. Its HSS T3 particles are designed for separations requiring silica-based selectivities or increased retention of hydrophilic peptides. It is specifically designed to protect 2.1 mm ID analytical columns.

  • Pore size: 100 Å
  • Particle size: 2.5 μm
  • Inner diameter: 2.1 mm
  • Length: 5 mm
  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8
  • Temperature limits: 45 °C
  • Maximum pressure: 18,000 psi (1240 bar)
  • Molecular weight range: 1000 to 15000
  • System: UHPLC
  • USP classification: L1
  • Units per package: 3 pk

Catalog No. 50-239-1861 Supplier Waters Corp Supplier No. 186008761
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This guard column is optimized to protect and prolong analytical column lifetimes without compromising chromatographic performance. Its HSS T3 particles are designed for separations requiring silica-based selectivities or increased retention of hydrophilic peptides. It is specifically designed to protect 2.1 mm ID analytical columns.

  • Pore size: 100 Å
  • Particle size: 2.5 μm
  • Inner diameter: 2.1 mm
  • Length: 5 mm
  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8
  • Temperature limits: 45 °C
  • Maximum pressure: 18,000 psi (1240 bar)
  • Molecular weight range: 1000 to 15000
  • System: UHPLC
  • USP classification: L1
  • Units per package: 3 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.