Learn More
Waters Corp XSelect Peptide HSS T3 Column, 100à , 5 µm, 4.6 mm X 100 mm, 1K - 15K, 1/pk
Catalog No. 502391998 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect Peptide HSS T3 Column, 100à , 5 µm, 4.6 mm X 100 mm, 1K - 15K, 1/pk
Catalog No. 50-239-1998 Supplier Waters Corp Supplier No. 186008779
XSelect Peptide HSS T3 Column, 100à , 5 µm, 4.6 mm X 100 mm, 1K - 15K, 1/pk