Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp XSelect Peptide HSS T3 Column, 100 Å, 5 μm

Catalog No. 502391850 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The XSelect Peptide HSS T3 Column is engineered to provide increased peptide retentivity and unique selectivity. This column features a 100 Å pore size and 5 μm particle size, with dimensions of 4.6 x 250 mm, making it suitable for various chromatographic applications.

  • Manufactured in a cGMP, ISO 9001:2000 certified plant.
  • Each batch is chromatographically tested with acidic, basic, and neutral analytes.
  • QC tested with a gradient separation of a tryptic digest of cytochrome c.
  • Equipped with style endfittings.
  • Withstands maximum pressure up to 5000 psi (400 bar or 40 Mpa).
  • Recommended operating pH range is 2 to 8.
  • Maximum recommended temperature is 45 °C.
  • Shipped in 100% acetonitrile solvent.

Catalog No. 50-239-1850 Supplier Waters Corp Supplier No. 176004042
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The XSelect Peptide HSS T3 Column is engineered to provide increased peptide retentivity and unique selectivity. This column features a 100 Å pore size and 5 μm particle size, with dimensions of 4.6 x 250 mm, making it suitable for various chromatographic applications.

  • Manufactured in a cGMP, ISO 9001:2000 certified plant.
  • Each batch is chromatographically tested with acidic, basic, and neutral analytes.
  • QC tested with a gradient separation of a tryptic digest of cytochrome c.
  • Equipped with style endfittings.
  • Withstands maximum pressure up to 5000 psi (400 bar or 40 Mpa).
  • Recommended operating pH range is 2 to 8.
  • Maximum recommended temperature is 45 °C.
  • Shipped in 100% acetonitrile solvent.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.