Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp XSelect Peptide HSS T3 Column, 100Å, 5 μm

Catalog No. 502392707 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

XSelect Peptide HSS T3 Columns are designed for chromatographic separations requiring silica-based selectivities and increased retention of hydrophilic peptides. Each batch of HSS T3, 100Å material undergoes rigorous Quality Control (QC) testing to ensure stringent performance criteria are met. This Method Validation Kit (MVK) includes three columns from three different material batches, ensuring consistency and reliability.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8
  • Temperature limits: 45°C
  • Maximum pressure: 6000 psi (415 Bar)
  • Bonding technology: T3
  • Molecular weight range: 1000 to 15000
  • Particle size: 5 μm
  • Pore size: 100 Å
  • Inner diameter: 4.6 mm
  • Length: 150 mm
  • System: HPLC
  • Units per package: 3

Catalog No. 50-239-2707 Supplier Waters Corp Supplier No. 186008788
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

XSelect Peptide HSS T3 Columns are designed for chromatographic separations requiring silica-based selectivities and increased retention of hydrophilic peptides. Each batch of HSS T3, 100Å material undergoes rigorous Quality Control (QC) testing to ensure stringent performance criteria are met. This Method Validation Kit (MVK) includes three columns from three different material batches, ensuring consistency and reliability.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8
  • Temperature limits: 45°C
  • Maximum pressure: 6000 psi (415 Bar)
  • Bonding technology: T3
  • Molecular weight range: 1000 to 15000
  • Particle size: 5 μm
  • Pore size: 100 Å
  • Inner diameter: 4.6 mm
  • Length: 150 mm
  • System: HPLC
  • Units per package: 3

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.