Learn More
Waters Corp XSelect Peptide HSS T3 Column, 100 Å, 5 μm, 2.1 x 150 mm
Supplier: Waters Corp 176004018
This kit contains a high-performance C18 column designed for peptide separations in HPLC systems. It features C18 chemistry on a silica particle substrate with T3 bonding technology, offering broad pH stability and high-pressure capabilities. The included Cytochrome C Digestion Standard enables immediate performance benchmarking and continuous monitoring.
- C18 chemistry for reversed phase separation.
- Silica particle substrate.
- pH range of 2 to 8.
- Temperature limits up to 45°C.
- Maximum pressure of 6000 psi (415 Bar).
- T3 bonding technology.
- Spherical particle shape with a size of 5 μm.
- Pore size of 100 Å.
- Quality control tested for peptide applications.
- Inner diameter of 2.1 mm and length of 150 mm.
- Includes a Cytochrome C Digestion Standard for performance monitoring.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.