Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp XSelect Column, C18, 100 Å, 5 μm, 4.6 × 150 mm

Catalog No. 502390486 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This kit includes one Peptide HSS T3 column and a Cytochrome C Digestion Standard vial for immediate benchmarking. Reorderable for continuous monitoring, each batch is prepared under strict quality control and confirmed by a certificate of analysis.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • PH range: 2–8
  • Temperature limit: 45 °C
  • Max pressure: 6000 psi (415 Bar)
  • Endcapped: Yes
  • Bonding technology: T3
  • Silanol activity: Medium
  • Molecular weight range: 1000–15000
  • Particle shape: Spherical
  • Particle size: 5 μm
  • Pore size: 100 Å
  • QC tested: Peptide
  • Surface area: 230
  • Particle technology: HSS
  • USP classification: L1
  • ID: 4.6 mm
  • Length: 150 mm
  • Carbon load: 11%
  • Application: Peptide
  • Units per package: 1

Catalog No. 50-239-0486 Supplier Waters Corp Supplier No. 176004021
May include imposed supplier surcharges.
Add to Cart
Add to Cart

This kit includes one Peptide HSS T3 column and a Cytochrome C Digestion Standard vial for immediate benchmarking. Reorderable for continuous monitoring, each batch is prepared under strict quality control and confirmed by a certificate of analysis.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • PH range: 2–8
  • Temperature limit: 45 °C
  • Max pressure: 6000 psi (415 Bar)
  • Endcapped: Yes
  • Bonding technology: T3
  • Silanol activity: Medium
  • Molecular weight range: 1000–15000
  • Particle shape: Spherical
  • Particle size: 5 μm
  • Pore size: 100 Å
  • QC tested: Peptide
  • Surface area: 230
  • Particle technology: HSS
  • USP classification: L1
  • ID: 4.6 mm
  • Length: 150 mm
  • Carbon load: 11%
  • Application: Peptide
  • Units per package: 1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.