Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Guard Column, 2.5 μm, 3.9 mm ID, 5 mm L, 100 Å

Catalog No. 502391866 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This guard column protects and extends the lifetime of analytical columns while maintaining chromatographic performance. It features HSS T3 particles for silica-based selectivities or increased retention of hydrophilic peptides. This cartridge is designed to protect a 4.6 mm ID analytical column and requires a reusable universal guard cartridge holder.

  • Protects and extends analytical column lifetime.
  • Maintains chromatographic performance.
  • Features HSS T3 particles for silica-based selectivities.
  • Increases retention of hydrophilic peptides.
  • Particle size: 2.5 μm.
  • Pore size: 100 Å.
  • Dimensions: 3.9 mm ID x 5 mm L.
  • Chemistry: C18, Reversed Phase.
  • pH range: 2 to 8.
  • Temperature limits: 45 °C.
  • Compatible with UPLC, UHPLC, and HPLC systems.
  • Units per package: 3.

Catalog No. 50-239-1866 Supplier Waters Corp Supplier No. 186008765
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This guard column protects and extends the lifetime of analytical columns while maintaining chromatographic performance. It features HSS T3 particles for silica-based selectivities or increased retention of hydrophilic peptides. This cartridge is designed to protect a 4.6 mm ID analytical column and requires a reusable universal guard cartridge holder.

  • Protects and extends analytical column lifetime.
  • Maintains chromatographic performance.
  • Features HSS T3 particles for silica-based selectivities.
  • Increases retention of hydrophilic peptides.
  • Particle size: 2.5 μm.
  • Pore size: 100 Å.
  • Dimensions: 3.9 mm ID x 5 mm L.
  • Chemistry: C18, Reversed Phase.
  • pH range: 2 to 8.
  • Temperature limits: 45 °C.
  • Compatible with UPLC, UHPLC, and HPLC systems.
  • Units per package: 3.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.