Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp XSelect Peptide HSS T3 VanGuard Cartridge, 100Å, 1.7 μm, 2.1 mm × 5 mm

Catalog No. 502391995 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

XSelect Peptide HSS T3 VanGuard Cartridges protect and extend the lifetime of analytical columns, maintaining chromatographic performance. They feature HSS T3 particles, ideal for silica-based selectivities or increased retention of hydrophilic peptides, and are designed to safeguard 2.1mm ID analytical columns.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8 pH
  • Maximum pressure: 18000 psi (1240 bar)
  • Endcapped: Yes
  • Bonding technology: T3
  • Particle size: 1.7 μm
  • Pore size: 100 Å
  • Format: VanGuard Cartridge
  • System compatibility: UPLC, UHPLC
  • Particle technology: HSS
  • Technique: LC
  • Inner diameter: 2.1 mm
  • Length: 5 mm
  • Carbon load: 11%
  • Application: Peptide

Catalog No. 50-239-1995 Supplier Waters Corp Supplier No. 186008777
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

XSelect Peptide HSS T3 VanGuard Cartridges protect and extend the lifetime of analytical columns, maintaining chromatographic performance. They feature HSS T3 particles, ideal for silica-based selectivities or increased retention of hydrophilic peptides, and are designed to safeguard 2.1mm ID analytical columns.

  • Chemistry: C18
  • Separation mode: Reversed phase
  • Particle substrate: Silica
  • pH range: 2 to 8 pH
  • Maximum pressure: 18000 psi (1240 bar)
  • Endcapped: Yes
  • Bonding technology: T3
  • Particle size: 1.7 μm
  • Pore size: 100 Å
  • Format: VanGuard Cartridge
  • System compatibility: UPLC, UHPLC
  • Particle technology: HSS
  • Technique: LC
  • Inner diameter: 2.1 mm
  • Length: 5 mm
  • Carbon load: 11%
  • Application: Peptide

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.