Learn More
Waters Corp WATERS CORPORATION XSelect Analytical Column, Phenyl-Hexyl, 3.0 x 75 mm, 3.5 μm
Catalog No. 50466178 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
WATERS CORPORATION XSelect Analytical Column, Phenyl-Hexyl, 3.0 x 75 mm, 3.5 μm
Catalog No. 50-466-178 Supplier Waters Corp Supplier No. 186005648
WATERS CORPORATION XSelect Analytical Column, Phenyl-Hexyl, 3.0 x 75 mm, 3.5 μm