Learn More
Waters Corp XSelect Premier CSH C18 Column, 3.5 μm, 4.6 mm ID, 100 mm Length
Catalog No. 502886208 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect Premier CSH C18 Columns are general purpose C18 columns providing alternative selectivity. They feature Charged Surface Hybrid (CSH) technology for superior peak shape and increased loading capacity for basic compounds. These columns offer exceptional separation performance, robustness, and throughput for HPLC assays, facilitating a seamless transition to UPLC. MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • Charged Surface Hybrid (CSH) technology for superior peak shape and increased loading capacity for basic compounds.
  • Exceptional separation performance, robustness, and throughput for HPLC assays.
  • Seamless transition path toward UPLC adoption.
  • MaxPeak High Performance Surface hardware technology to reduce unwanted analyte/surface interactions.
Catalog No. 50-288-6208 Supplier Waters Corp Supplier No. 186010643
XSelect Premier CSH C18 Columns are general purpose C18 columns providing alternative selectivity. They feature Charged Surface Hybrid (CSH) technology for superior peak shape and increased loading capacity for basic compounds. These columns offer exceptional separation performance, robustness, and throughput for HPLC assays, facilitating a seamless transition to UPLC. MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • Charged Surface Hybrid (CSH) technology for superior peak shape and increased loading capacity for basic compounds.
  • Exceptional separation performance, robustness, and throughput for HPLC assays.
  • Seamless transition path toward UPLC adoption.
  • MaxPeak High Performance Surface hardware technology to reduce unwanted analyte/surface interactions.