Learn More
Waters Corp XSelect Premier CSH C18 Column, 3.5 μm, 2.1 x 100 mm, 130 Å
Catalog No. 502886214 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect Premier CSH C18 Column is engineered for high-performance bioseparations, providing enhanced low-level detection of impurities and excellent mass spectrometry performance for various analytical applications.
  • Accepts greater peptide mass loads for improved low-level detection of impurities.
  • Offers excellent performance with TFA for optical applications, FA for MS, and DFA for dual detection.
  • Provides excellent peak shape for basic compounds under acidic, low ionic strength conditions.
  • Delivers excellent MS performance with formic acid as a mobile phase modifier.
  • Enables fast pH switching and column equilibration.
Catalog No. 50-288-6214 Supplier Waters Corp Supplier No. 186010631
The XSelect Premier CSH C18 Column is engineered for high-performance bioseparations, providing enhanced low-level detection of impurities and excellent mass spectrometry performance for various analytical applications.
  • Accepts greater peptide mass loads for improved low-level detection of impurities.
  • Offers excellent performance with TFA for optical applications, FA for MS, and DFA for dual detection.
  • Provides excellent peak shape for basic compounds under acidic, low ionic strength conditions.
  • Delivers excellent MS performance with formic acid as a mobile phase modifier.
  • Enables fast pH switching and column equilibration.