Learn More
Waters Corp Column, XSelect Premier CSH C18, 3.5 μm, 4.6 x 150 mm
Catalog No. 502886254 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XSelect Premier CSH C18 Columns are C18 columns featuring Charged Surface Hybrid (CSH) and MaxPeak High Performance Surface (HPS) technologies for enhanced HPLC assay performance.
  • CSH technology provides superior peak shape and increased loading capacity for basic compounds.
  • MaxPeak HPS technology reduces unwanted analyte/surface interactions, enhancing recovery, sensitivity, and reproducibility.
  • Ensures exceptional separation performance, robustness, and throughput for HPLC assays.
  • Allows for fast pH switching, column equilibration, and reduced conditioning times.
Catalog No. 50-288-6254 Supplier Waters Corp Supplier No. 186010644
XSelect Premier CSH C18 Columns are C18 columns featuring Charged Surface Hybrid (CSH) and MaxPeak High Performance Surface (HPS) technologies for enhanced HPLC assay performance.
  • CSH technology provides superior peak shape and increased loading capacity for basic compounds.
  • MaxPeak HPS technology reduces unwanted analyte/surface interactions, enhancing recovery, sensitivity, and reproducibility.
  • Ensures exceptional separation performance, robustness, and throughput for HPLC assays.
  • Allows for fast pH switching, column equilibration, and reduced conditioning times.