Learn More
Waters Corp VanGuard FIT Column, 3.5 μm, 2.1 x 50 mm, 130 Å
Supplier:  Waters Corp 186010626
Encompass_Preferred
The XSelect Premier CSH C18 VanGuard FIT Column is designed with a particle size of 3.5 μm, 2.1 x 50 mm dimensions, and a 130 Å pore diameter. This column provides excellent peak shape for basic compounds, good MS performance with formic acid, and enables fast pH switching and column equilibration.
  • Excellent peak shape for basic compounds under acidic, low ionic strength conditions.
  • Excellent MS performance with formic acid as a mobile phase modifier.
  • Fast pH switching and column equilibration.
Catalog No. 50-288-6198