Learn More
Waters Corp VanGuard FIT Cartridge, CSH C18, 3.5 μm, 2.1 x 5 mm
Catalog No. 502886259 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This guard cartridge protects analytical columns from contamination, extending life and ensuring stable HPLC performance. It features a 3.5 μm particle size and 2.1 x 5 mm dimensions.
  • Protects analytical column from contamination
  • Extends column lifetime
  • Provides stable HPLC performance
  • 3.5 μm particle size
  • 2.1 x 5 mm dimensions
  • 3-pack configuration
  • C18 chemistry
  • pH range 1 to 11
  • Maximum pressure 6000 psi (415 Bar)
  • Pore size 130 Å
  • HPLC system compatible
  • Guard format
Catalog No. 50-288-6259 Supplier Waters Corp Supplier No. 186010629
This guard cartridge protects analytical columns from contamination, extending life and ensuring stable HPLC performance. It features a 3.5 μm particle size and 2.1 x 5 mm dimensions.
  • Protects analytical column from contamination
  • Extends column lifetime
  • Provides stable HPLC performance
  • 3.5 μm particle size
  • 2.1 x 5 mm dimensions
  • 3-pack configuration
  • C18 chemistry
  • pH range 1 to 11
  • Maximum pressure 6000 psi (415 Bar)
  • Pore size 130 Å
  • HPLC system compatible
  • Guard format