Learn More
Waters Corp Column, CSH C18, 3.5 μm, 4.6 x 100 mm
Catalog No. 502886260 Shop All Waters Corporation Products
Encompass_Preferred
The XSelect Premier CSH C18 VanGuard FIT Column, with MaxPeak High Performance Surfaces (HPS), enhances analyte recovery, sensitivity, and reproducibility by minimizing analyte/surface interactions. CSH™ Technology ensures excellent peak shape for basic compounds, superior MS performance with formic acid, and fast pH switching and column equilibration.
  • Reduced column conditioning and passivation times.
  • Improved sensitivity and peak shapes.
  • Improved recovery.
  • Simpler mobile phases, without complex additives.
  • Time savings in method development.
  • Reduced risk and greater confidence in data and decision making.
  • The “VanGuard FIT” designation indicates that the column integrates a guard column to maintain separation efficiency and extend column lifetime.
Catalog No. 50-288-6260 Supplier Waters Corp Supplier No. 186010637
The XSelect Premier CSH C18 VanGuard FIT Column, with MaxPeak High Performance Surfaces (HPS), enhances analyte recovery, sensitivity, and reproducibility by minimizing analyte/surface interactions. CSH™ Technology ensures excellent peak shape for basic compounds, superior MS performance with formic acid, and fast pH switching and column equilibration.
  • Reduced column conditioning and passivation times.
  • Improved sensitivity and peak shapes.
  • Improved recovery.
  • Simpler mobile phases, without complex additives.
  • Time savings in method development.
  • Reduced risk and greater confidence in data and decision making.
  • The “VanGuard FIT” designation indicates that the column integrates a guard column to maintain separation efficiency and extend column lifetime.