Learn More
Waters Corp Column, CSH C18, 3.5 μm, 4.6 x 150 mm
Catalog No. 502886224 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These C18 columns leverage Charged Surface Hybrid (CSH) technology for superior peak shape and increased loading capacity of basic compounds. Designed for exceptional separation performance, robustness, and throughput in HPLC assays, they facilitate a seamless transition to UPLC. MaxPeak High Performance Surface hardware minimizes unwanted analyte/surface interactions.
  • General purpose C18 columns provide alternative selectivity compared to other C18 sorbents.
  • Charged Surface Hybrid (CSH) technology provides superior peak shape and increased loading capacity for basic compounds.
  • Exceptional separation performance, robustness, and throughput for HPLC assays.
  • Seamless transition path toward UPLC adoption.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions, which can lead to poor peak shape and signal intensity losses.
Catalog No. 50-288-6224 Supplier Waters Corp Supplier No. 186010639
These C18 columns leverage Charged Surface Hybrid (CSH) technology for superior peak shape and increased loading capacity of basic compounds. Designed for exceptional separation performance, robustness, and throughput in HPLC assays, they facilitate a seamless transition to UPLC. MaxPeak High Performance Surface hardware minimizes unwanted analyte/surface interactions.
  • General purpose C18 columns provide alternative selectivity compared to other C18 sorbents.
  • Charged Surface Hybrid (CSH) technology provides superior peak shape and increased loading capacity for basic compounds.
  • Exceptional separation performance, robustness, and throughput for HPLC assays.
  • Seamless transition path toward UPLC adoption.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions, which can lead to poor peak shape and signal intensity losses.