Learn More
Waters Corp Waters Column, Phenyl-Hexyl, 2.5 μm, 2.1 x 50 mm, 130 Å
Supplier:  Waters Corp 186009879
Encompass_Preferred
These columns provide alternative analyte selectivity, utilizing a robust C6 phenyl-ligand and MaxPeak High Performance Surface technology. They enhance separation performance and throughput for HPLC and UPLC assays by minimizing unwanted analyte/surface interactions.
  • Offers alternative analyte selectivity and unique retention for polyaromatic compounds.
  • Robust, low bleed C6 phenyl-ligand for selective pi-pi interactions.
  • MaxPeak HPS technology reduces unwanted analyte/surface interactions.
  • Enhances separation performance, robustness, and throughput.
  • Improves analyte recovery, sensitivity, and peak shapes.
  • Shortens column conditioning and passivation times.
  • Ensures excellent peak shape for basic compounds and MS performance.
Catalog No. 50-222-4296