Learn More
Waters Corp Waters XSelect Premier CSH Column, 130Å, 2.5 μm, 2.1 × 150 mm
Supplier:  Waters Corp 186009881
Encompass_Preferred
These columns provide alternative analyte selectivity, crucial for method development. Their trifunctionally bonded C6 phenyl-ligand ensures a robust, low bleed sorbent that selectively retains polyaromatic compounds through π–π interactions. MaxPeak High Performance Surface hardware technology minimizes unwanted analyte/surface interactions, preventing poor peak shape and signal intensity reductions.
  • Alternative analyte selectivity for method development.
  • Robust, low bleed C6 phenyl-ligand for selective retention of polyaromatic compounds.
  • Exceptional separation performance and throughput for HPLC assays.
  • Seamless transition to UPLC adoption.
  • MaxPeak High Performance Surface hardware minimizes unwanted analyte/surface interactions.
  • Reduces poor peak shape and prevents signal intensity loss.
  • Digital access to column documents at www.waters.com/coa.
Catalog No. 50-222-4298