Learn More
Waters Corp Column, HSS C18, 2.5 μm, 2.1 mm x 150 mm
Catalog No. 502391114 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These XSelect Premier HSS C18 Columns utilize a C18 bonded phase for increased retention and provide long lifetimes at low pH due to HSS particles. MaxPeak High Performance Surface minimizes unwanted analyte/surface interactions, improving peak shape and signal. The integrated VanGuard Fit design protects against contaminants, extending column longevity.
  • Increased retention from C18 bonded phase.
  • Long column lifetimes at low pH with HSS particle technology.
  • MaxPeak HPS minimizes analyte/surface interactions for better peak shape and signal.
  • VanGuard Fit design extends column lifetime and protects against contaminants.
  • Enhances analyte recovery, sensitivity, and reproducibility.
Catalog No. 50-239-1114 Supplier Waters Corp Supplier No. 186010027
These XSelect Premier HSS C18 Columns utilize a C18 bonded phase for increased retention and provide long lifetimes at low pH due to HSS particles. MaxPeak High Performance Surface minimizes unwanted analyte/surface interactions, improving peak shape and signal. The integrated VanGuard Fit design protects against contaminants, extending column longevity.
  • Increased retention from C18 bonded phase.
  • Long column lifetimes at low pH with HSS particle technology.
  • MaxPeak HPS minimizes analyte/surface interactions for better peak shape and signal.
  • VanGuard Fit design extends column lifetime and protects against contaminants.
  • Enhances analyte recovery, sensitivity, and reproducibility.