Learn More
Waters Corp Column, HSS C18, VanGuard FIT, 2.5 μm, 4.6 mm x 100 mm
Catalog No. 502392001 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
  • Ultra-performance general purpose C18 bonded phase provides increased retention.
  • High Strength Silica (HSS) particle and tri-functional C18 bonded phase ensure extremely long column lifetimes at low pH.
  • MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • VanGuard Fit column design protects the column from contaminants and extends column lifetime.
Catalog No. 50-239-2001 Supplier Waters Corp Supplier No. 186010033
  • Ultra-performance general purpose C18 bonded phase provides increased retention.
  • High Strength Silica (HSS) particle and tri-functional C18 bonded phase ensure extremely long column lifetimes at low pH.
  • MaxPeak High Performance Surface hardware technology significantly reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • VanGuard Fit column design protects the column from contaminants and extends column lifetime.