Learn More
Waters Corp Column, XSelect Premier HSS C18, 2.5 μm, 4.6 mm ID, 50 mm Length, MaxPeak HPS
Catalog No. 502392810 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect Premier HSS C18 Columns are ultra-performance general-purpose C18 bonded phase columns offering increased retention. They feature high strength silica (HSS) particles and a tri-functional C18 bonded phase for exceptionally long column lifetimes at low pH. MaxPeak High Performance Surface hardware minimizes undesirable analyte/surface interactions, improving peak shape and signal intensity. The VanGuard Fit column design protects against contaminants, extending column lifetime.
  • MaxPeak High Performance Surface hardware reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • VanGuard Fit column design protects from contaminants and increases column lifetime.
  • High strength silica (HSS) particles and tri-functional C18 bonded phase ensure extended column lifetimes at low pH.
  • Ultra-performance general-purpose C18 bonded phase provides increased retention.
Catalog No. 50-239-2810 Supplier Waters Corp Supplier No. 186010032
The XSelect Premier HSS C18 Columns are ultra-performance general-purpose C18 bonded phase columns offering increased retention. They feature high strength silica (HSS) particles and a tri-functional C18 bonded phase for exceptionally long column lifetimes at low pH. MaxPeak High Performance Surface hardware minimizes undesirable analyte/surface interactions, improving peak shape and signal intensity. The VanGuard Fit column design protects against contaminants, extending column lifetime.
  • MaxPeak High Performance Surface hardware reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • VanGuard Fit column design protects from contaminants and increases column lifetime.
  • High strength silica (HSS) particles and tri-functional C18 bonded phase ensure extended column lifetimes at low pH.
  • Ultra-performance general-purpose C18 bonded phase provides increased retention.