Learn More
Waters Corp Column, PFP, 2.5 μm, 4.6 mm, 100 mm
Catalog No. 502392312 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Premier HSS PFP Column is a non-endcapped, silica-based column engineered for low pH separations, providing alternative selectivity to fully endcapped C18 phases. It incorporates MaxPeak High Performance Surface hardware technology to significantly reduce unwanted analyte/surface interactions, which often cause poor peak shape and decreased signal intensity.
  • HSS Technology provides increased retentivity and mechanical stability.
  • MaxPeak High Performance Surfaces minimize analyte/surface interactions, enhancing recovery and sensitivity.
  • Mitigates loss of metal-sensitive analytes.
  • Reduces column conditioning and improves peak shapes.
  • Features PFP/fluoro-phenyl chemistry for reversed-phase.
  • Dimensions: 2.5 μm particle size, 4.6 mm ID, 100 mm length.
  • pH range: 2 to 8; Max pressure: 18000 psi (1240 Bar).
  • Compatible with UHPLC and UPLC systems.
Catalog No. 50-239-2312 Supplier Waters Corp Supplier No. 186010051
The Premier HSS PFP Column is a non-endcapped, silica-based column engineered for low pH separations, providing alternative selectivity to fully endcapped C18 phases. It incorporates MaxPeak High Performance Surface hardware technology to significantly reduce unwanted analyte/surface interactions, which often cause poor peak shape and decreased signal intensity.
  • HSS Technology provides increased retentivity and mechanical stability.
  • MaxPeak High Performance Surfaces minimize analyte/surface interactions, enhancing recovery and sensitivity.
  • Mitigates loss of metal-sensitive analytes.
  • Reduces column conditioning and improves peak shapes.
  • Features PFP/fluoro-phenyl chemistry for reversed-phase.
  • Dimensions: 2.5 μm particle size, 4.6 mm ID, 100 mm length.
  • pH range: 2 to 8; Max pressure: 18000 psi (1240 Bar).
  • Compatible with UHPLC and UPLC systems.