Learn More
Waters Corp XSelect Premier HSS PFP Column, 2.5 μm, 4.6 mm x 150 mm
Supplier:  Waters Corp 186010052
Encompass_Preferred
This non-endcapped, silica-based column is designed for low pH separations that require alternative selectivity compared to fully endcapped C18 phases. It incorporates advanced surface technology to significantly reduce unwanted analyte/surface interactions, improving chromatographic performance.
  • Designed for low pH separations requiring alternative selectivity.
  • Non-endcapped, silica-based column.
  • Advanced surface technology reduces unwanted analyte/surface interactions.
  • Minimizes poor peak shape.
  • Prevents losses in signal intensity.
Catalog No. 50-239-0757