Learn More
Waters Corp VanGuard FIT Cartridge, 2.5 μm, 2.1 x 5 mm, 3/pk
Catalog No. 502391920 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect Premier HSS PFP VanGuard FIT Cartridge features MaxPeak High Performance Surfaces (HPS) technology to minimize analyte interactions, enhancing sensitivity, peak shapes, and recovery. Its high strength silica (HSS) provides mechanical stability and increased retentivity, protecting the main column and extending its lifespan.
  • Reduced column conditioning and passivation times.
  • Improved sensitivity, peak shapes, and recovery.
  • Simpler mobile phases, without complex additives.
  • Mitigates the loss of metal-sensitive analytes (e.g., lipids, acidic peptides).
  • Increased retentivity over hybrid materials.
  • Integrated guard column technology for maintained separation efficiency.
  • Extends column lifetime.
Catalog No. 50-239-1920 Supplier Waters Corp Supplier No. 186010049
The XSelect Premier HSS PFP VanGuard FIT Cartridge features MaxPeak High Performance Surfaces (HPS) technology to minimize analyte interactions, enhancing sensitivity, peak shapes, and recovery. Its high strength silica (HSS) provides mechanical stability and increased retentivity, protecting the main column and extending its lifespan.
  • Reduced column conditioning and passivation times.
  • Improved sensitivity, peak shapes, and recovery.
  • Simpler mobile phases, without complex additives.
  • Mitigates the loss of metal-sensitive analytes (e.g., lipids, acidic peptides).
  • Increased retentivity over hybrid materials.
  • Integrated guard column technology for maintained separation efficiency.
  • Extends column lifetime.