Learn More
Waters Corp VanGuard FIT Cartridge, XSelect Premier HSS PFP, 100 Å, 2.5 μm
Supplier:  Waters Corp 186010056
Encompass_Preferred
This VanGuard FIT Cartridge is designed for XSelect Premier HSS PFP 100 Å, 2.5 μm columns. It has dimensions of 3.9 x 5 mm.
  • Designed to protect your column investment.
  • Integrates a guard column to maintain separation efficiency.
  • Extends column lifetime.
Catalog No. 50-239-2298