Learn More
Waters Corp Waters VanGuard FIT Column, 2.5 μm, 4.6 mm x 100 mm, HSS PFP
Catalog No. 502392386 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This VanGuard FIT Column from the XSelect Premier HSS PFP series features MaxPeak High Performance Surfaces. It integrates a guard column to maintain separation efficiency and extend column lifetime.
  • Reduced column conditioning and passivation times
  • Improved sensitivity and peak shapes
  • Simpler mobile phases, without complex additives
  • Time savings in method development
  • Reduced risk and greater confidence in data and decision making
  • Increased retentivity over hybrid materials
  • Widest selectivity space with C18, T3, C18 SB, Cyano, and PFP chemistries
  • High strength silica (HSS) for mechanical stability
Catalog No. 50-239-2386 Supplier Waters Corp Supplier No. 186010054
This VanGuard FIT Column from the XSelect Premier HSS PFP series features MaxPeak High Performance Surfaces. It integrates a guard column to maintain separation efficiency and extend column lifetime.
  • Reduced column conditioning and passivation times
  • Improved sensitivity and peak shapes
  • Simpler mobile phases, without complex additives
  • Time savings in method development
  • Reduced risk and greater confidence in data and decision making
  • Increased retentivity over hybrid materials
  • Widest selectivity space with C18, T3, C18 SB, Cyano, and PFP chemistries
  • High strength silica (HSS) for mechanical stability