Learn More
Waters Corp Column, XSelect Premier HSS PFP VanGuard FIT, 2.5 μm, 2.1 mm x 150 mm
Catalog No. 502390821 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect Premier HSS PFP Column is a non-endcapped silica-based column designed for low pH separations that require alternative selectivity compared to a fully endcapped, high coverage C18 phase. It incorporates MaxPeak High Performance Surface hardware technology, which significantly reduces unwanted analyte/surface interactions that can lead to poor peak shape and losses in signal intensity. The VanGuard Fit column design serves to protect the column from contaminants and extends its lifetime.
  • Designed for low pH separations.
  • Offers alternative selectivity compared to fully endcapped, high coverage C18 phases.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • VanGuard Fit column design protects against contaminants and increases column lifetime.
Catalog No. 50-239-0821 Supplier Waters Corp Supplier No. 186010048
Add to Cart
Add to Cart
The XSelect Premier HSS PFP Column is a non-endcapped silica-based column designed for low pH separations that require alternative selectivity compared to a fully endcapped, high coverage C18 phase. It incorporates MaxPeak High Performance Surface hardware technology, which significantly reduces unwanted analyte/surface interactions that can lead to poor peak shape and losses in signal intensity. The VanGuard Fit column design serves to protect the column from contaminants and extends its lifetime.
  • Designed for low pH separations.
  • Offers alternative selectivity compared to fully endcapped, high coverage C18 phases.
  • MaxPeak High Performance Surface hardware technology reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • VanGuard Fit column design protects against contaminants and increases column lifetime.