Learn More
Waters Corp WATERS CORPORATION XSelect Premier HSS T3 Column, 100Å, 2.5 μm, 4.6 mm, 150 mm
Catalog No. 502224278 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XSelect Premier HSS T3 is a low-coverage C18 phase, compatible with 100% aqueous mobile phase for polar and non-polar compound separations. It features ultra-low MS bleed, is fully scalable to UPLC separations, and utilizes MaxPeak High Performance Surface hardware to reduce unwanted analyte interactions and signal loss.
  • MaxPeak High Performance Surface hardware technology for reduced analyte/surface interactions.
  • Offers exceptional separation performance, robustness, and throughput for UHPLC and HPLC assays.
  • Provides a seamless transition path to UPLC adoption.
Catalog No. 50-222-4278 Supplier Waters Corp Supplier No. 186009860
The XSelect Premier HSS T3 is a low-coverage C18 phase, compatible with 100% aqueous mobile phase for polar and non-polar compound separations. It features ultra-low MS bleed, is fully scalable to UPLC separations, and utilizes MaxPeak High Performance Surface hardware to reduce unwanted analyte interactions and signal loss.
  • MaxPeak High Performance Surface hardware technology for reduced analyte/surface interactions.
  • Offers exceptional separation performance, robustness, and throughput for UHPLC and HPLC assays.
  • Provides a seamless transition path to UPLC adoption.