Learn More
Waters Corp Premier Column, C18, 2.5 μm, 2.1 mm, 50 mm
Catalog No. NC3419586 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This C18 phase column is compatible with 100% aqueous mobile phase, ideal for separating polar and non-polar compounds. It exhibits ultra-low MS bleed, offering exceptional performance for UHPLC and HPLC assays. Integrated hardware technology reduces unwanted analyte/surface interactions, preventing poor peak shape and signal loss. Its design also protects from contaminants, extending column lifetime.
  • Low-coverage C18 phase compatible with 100% aqueous mobile phase for separating polar and non-polar compounds.
  • 2.5 μm columns enable exceptional separation performance, robustness and throughput for UHPLC and HPLC assays.
  • Hardware technology significantly reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • Column design protects from contaminants and increases column lifetime.
Catalog No. NC3419586 Supplier Waters Corp Supplier No. 1860098541EA
This C18 phase column is compatible with 100% aqueous mobile phase, ideal for separating polar and non-polar compounds. It exhibits ultra-low MS bleed, offering exceptional performance for UHPLC and HPLC assays. Integrated hardware technology reduces unwanted analyte/surface interactions, preventing poor peak shape and signal loss. Its design also protects from contaminants, extending column lifetime.
  • Low-coverage C18 phase compatible with 100% aqueous mobile phase for separating polar and non-polar compounds.
  • 2.5 μm columns enable exceptional separation performance, robustness and throughput for UHPLC and HPLC assays.
  • Hardware technology significantly reduces unwanted analyte/surface interactions, improving peak shape and signal intensity.
  • Column design protects from contaminants and increases column lifetime.