Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp XSelect Premier Peptide CSH C18 Column, 130 Å, 2.5 μm, 4.6 x 150 mm

Catalog No. 502392983 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This column is designed for peptide separations, featuring CSH C18 Column, 130 Å, 2.5 μm, 4.6 x 150 mm with Premier VanGuard FIT Guard particles. It provides excellent peak shape for peptides using FA- or TFA-containing mobile phases, is suitable for LC or LC-MS applications, and offers improved reproducibility, peak shape, and recovery for metal-sensitive analytes.

  • C18 chemistry, reversed phase, hybrid particle substrate
  • pH range: 1 to 11 pH; temperature limits: 80°C
  • Maximum pressure: 9000 psi (620 bar)
  • Particle size: 2.5 μm; pore size: 130 Å
  • Dimensions: 4.6 mm inner diameter, 150 mm length
  • Peptide QC tested, Premier column format
  • Compatible with UHPLC and UPLC systems for LC
  • Integrated, replaceable 5 mm pre-column
  • Ideal for peptide separations, analysis, mapping, and synthetic peptides

Catalog No. 50-239-2983 Supplier Waters Corp Supplier No. 186010717
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This column is designed for peptide separations, featuring CSH C18 Column, 130 Å, 2.5 μm, 4.6 x 150 mm with Premier VanGuard FIT Guard particles. It provides excellent peak shape for peptides using FA- or TFA-containing mobile phases, is suitable for LC or LC-MS applications, and offers improved reproducibility, peak shape, and recovery for metal-sensitive analytes.

  • C18 chemistry, reversed phase, hybrid particle substrate
  • pH range: 1 to 11 pH; temperature limits: 80°C
  • Maximum pressure: 9000 psi (620 bar)
  • Particle size: 2.5 μm; pore size: 130 Å
  • Dimensions: 4.6 mm inner diameter, 150 mm length
  • Peptide QC tested, Premier column format
  • Compatible with UHPLC and UPLC systems for LC
  • Integrated, replaceable 5 mm pre-column
  • Ideal for peptide separations, analysis, mapping, and synthetic peptides

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.