Learn More
Waters Corp VanGuard FIT Cartridge, 2.5 μm, C18, 3.9 x 5 mm, 3/pk
Catalog No. 502392984 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This VanGuard FIT Cartridge serves as a replacement for CSH C18 Columns with the Premier VanGuard FIT column design. Replace cartridges when high analytical column backpressure or reduced component resolution is evident.
  • C18 chemistry for reversed phase separation
  • Silica particle substrate, spherical shape, low silanol activity
  • pH range of 1 to 11; temperature limits up to 80°C
  • Maximum pressure of 18000 psi (1240 Bar)
  • 2.5 μm particle size, 130 Å pore size
  • Premier FIT Cartridge format with Parker-style endfitting
  • Peptide application suitability, QC tested for peptide
  • Compatible with UHPLC and UPLC systems
  • 3.9 mm inner diameter, 5 mm length
  • C18+ bonding and CSH particle technology
  • 15% carbon load; 185 surface area
  • Packed with acetonitrile; L1 USP classification
Catalog No. 50-239-2984 Supplier Waters Corp Supplier No. 186010719
This VanGuard FIT Cartridge serves as a replacement for CSH C18 Columns with the Premier VanGuard FIT column design. Replace cartridges when high analytical column backpressure or reduced component resolution is evident.
  • C18 chemistry for reversed phase separation
  • Silica particle substrate, spherical shape, low silanol activity
  • pH range of 1 to 11; temperature limits up to 80°C
  • Maximum pressure of 18000 psi (1240 Bar)
  • 2.5 μm particle size, 130 Å pore size
  • Premier FIT Cartridge format with Parker-style endfitting
  • Peptide application suitability, QC tested for peptide
  • Compatible with UHPLC and UPLC systems
  • 3.9 mm inner diameter, 5 mm length
  • C18+ bonding and CSH particle technology
  • 15% carbon load; 185 surface area
  • Packed with acetonitrile; L1 USP classification