Learn More
Waters Corp Column, Peptide CSH C18, 130 Å, 2.5 μm
Catalog No. 502390854 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
MaxPeak Premier Columns utilize MaxPeak High Performance Surfaces (HPS) technology to minimize analyte/surface interactions, enhancing recovery, sensitivity, and reproducibility in chromatographic separations. This technology mitigates the loss of metal-sensitive analytes like lipids, organic acids, and peptides. The Peptide CSH C18 column is tailored for peptide analysis and, with VanGuard FIT technology, extends column lifetime by integrating a guard column.
  • Reduced column conditioning and passivation times
  • Improved sensitivity and peak shapes
  • Improved recovery
  • Simpler mobile phases, without complex additives
  • Time savings in method development
  • Reduced risk and greater confidence in data and decision making
  • Maintained separation efficiency and extended column lifetime due to VanGuard FIT integrated guard column technology
Catalog No. 50-239-0854 Supplier Waters Corp Supplier No. 186010716
MaxPeak Premier Columns utilize MaxPeak High Performance Surfaces (HPS) technology to minimize analyte/surface interactions, enhancing recovery, sensitivity, and reproducibility in chromatographic separations. This technology mitigates the loss of metal-sensitive analytes like lipids, organic acids, and peptides. The Peptide CSH C18 column is tailored for peptide analysis and, with VanGuard FIT technology, extends column lifetime by integrating a guard column.
  • Reduced column conditioning and passivation times
  • Improved sensitivity and peak shapes
  • Improved recovery
  • Simpler mobile phases, without complex additives
  • Time savings in method development
  • Reduced risk and greater confidence in data and decision making
  • Maintained separation efficiency and extended column lifetime due to VanGuard FIT integrated guard column technology