Learn More
Waters Corp Column, Peptide CSH C18, 130 Å, 2.5 μm, 2.1 x 100 mm
Catalog No. 502391741 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters™ MaxPeak™ Premier Columns feature MaxPeak High Performance Surfaces (HPS) technology to enhance chromatographic separations. This reduces analyte/surface interactions, improving recovery, sensitivity, and reproducibility for metal-sensitive analytes. They are suitable for reversed-phase, HILIC, and SEC modes.
  • Reduced column conditioning and passivation times.
  • Improved sensitivity, peak shapes, and recovery.
  • Simpler mobile phases, reducing need for complex additives.
  • Outstanding peak capacity and superior peak shape for peptide analysis with TFA, DFA, FA.
  • Accepts greater peptide mass loads, improving low-level impurity detection.
  • Excellent performance with formic acid (FA) for Mass Spectrometry (MS).
  • Fast pH switching and equilibration.
Catalog No. 50-239-1741 Supplier Waters Corp Supplier No. 186010712
Waters™ MaxPeak™ Premier Columns feature MaxPeak High Performance Surfaces (HPS) technology to enhance chromatographic separations. This reduces analyte/surface interactions, improving recovery, sensitivity, and reproducibility for metal-sensitive analytes. They are suitable for reversed-phase, HILIC, and SEC modes.
  • Reduced column conditioning and passivation times.
  • Improved sensitivity, peak shapes, and recovery.
  • Simpler mobile phases, reducing need for complex additives.
  • Outstanding peak capacity and superior peak shape for peptide analysis with TFA, DFA, FA.
  • Accepts greater peptide mass loads, improving low-level impurity detection.
  • Excellent performance with formic acid (FA) for Mass Spectrometry (MS).
  • Fast pH switching and equilibration.